Assay Details
Gene Symbol: hspa4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199857.1 | NP_956151.1 | 10 - 11 | 1362 | 71 | |
GenBank mRNA | BC048063.1 | - | 10 - 11 | 1362 | 71 | |
BC065970.1 | - | 10 - 11 | 1391 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
