This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03136331_m1
See other ASAP2A GE Assays ›
Gene Symbol:
asap2a
Gene Name:
ArfGAP with SH3 domain, ankyrin repeat and PH domain 2a
Gene Aliases:
ddef2a, si:dkey-27p7.2
Chromosome Location:
Chr.17: 35097003 - 35225157 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03136331_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: asap2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045207.1 NP_001038672.1 21 - 22 2541 71
XM_005158734.4 XP_005158791.1 22 - 23 2571 71
XM_021467217.1 XP_021322892.1 23 - 24 2598 71
XM_021467218.1 XP_021322893.1 22 - 23 2589 71
XM_021467219.1 XP_021322894.1 23 - 24 2598 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of GTPase activity
GTPase activator activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0