This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03135462_m1
See other INTS11 GE Assays ›
Gene Symbol:
ints11
Gene Name:
integrator complex subunit 11
Gene Aliases:
cpsf3l, zgc:110671
Chromosome Location:
Chr.23: 37062157 - 37085959 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03135462_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ints11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001020621.1 NP_001018457.1 17 - 18 1843 94
XM_005166209.4 XP_005166266.1 16 - 17 2014 94
GenBank mRNA BC095364.1 - 17 - 18 1843 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

snRNA processing
regulation of RNA splicing
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0