This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03135191_g1
See other CCN2A GE Assays ›
Gene Symbol:
ccn2a
Gene Name:
cellular communication network factor 2a
Gene Aliases:
ctgf, ctgfa, hm:zeh1559, si:dkey-266k12.4, wu:fl25c03, zgc:136644
Chromosome Location:
Chr.20: 25340814 - 25343490 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03135191_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ccn2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001015041.2 NP_001015041.1 4 - 5 921 101
GenBank mRNA BC115209.1 - 4 - 5 921 101
KM655688.1 - 4 - 5 735 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
signal transduction
endoderm development
notochord development
regeneration
chordate embryonic development
definitive hemopoiesis
negative regulation of cell death
integrin binding
insulin-like growth factor binding
heparin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0