This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03133268_g1
See other FAT1A GE Assays ›
Gene Symbol:
fat1a
Gene Name:
FAT atypical cadherin 1a
Gene Aliases:
fat, fat1
Chromosome Location:
Chr.1: 190965 - 346104 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03133268_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fat1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212967.2 NP_998132.2 16 - 17 10324 88
XM_005168343.4 XP_005168400.1 17 - 18 10438 88
XM_005168345.4 XP_005168402.1 17 - 18 10438 88
XM_021469206.1 XP_021324881.1 17 - 18 10438 88
XM_021469232.1 XP_021324907.1 17 - 18 10547 88
XM_021469270.1 XP_021324945.1 16 - 17 10369 88
GenBank mRNA AF469608.1 - 16 - 17 10322 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
pronephros development
cell-cell adhesion
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0