This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03128899_g1
See other PPP6R2A GE Assays ›
Gene Symbol:
ppp6r2a
Gene Name:
protein phosphatase 6, regulatory subunit 2a
Gene Aliases:
fd18a05, ppp6r2, saps2, si:dkey-159a18.3, wu:fd18a05, zgc:136621
Chromosome Location:
Chr.4: 20082475 - 20101739 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03128899_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ppp6r2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025509.1 NP_001020680.1 12 - 13 1989 69
XM_021475043.1 XP_021330718.1 11 - 12 1782 69
GenBank mRNA BC115199.1 - 12 - 13 1873 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of phosphoprotein phosphatase activity
protein phosphatase regulator activity
protein phosphatase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0