This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03128386_g1
See other RBM39B GE Assays ›
Gene Symbol:
rbm39b
Gene Name:
RNA binding motif protein 39b
Gene Aliases:
rnpc2l, wu:fa97g07, wu:fb09c08, zgc:112139, zgc:113117
Chromosome Location:
Chr.8: 37231752 - 37249832 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03128386_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rbm39b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001014370.1 NP_001014392.1 4 - 5 399 67
XM_005167222.4 XP_005167279.1 3 - 4 349 67
XM_005167224.4 XP_005167281.1 4 - 5 424 67
XM_009304133.3 XP_009302408.1 5 - 6 570 67
XM_021478249.1 XP_021333924.1 3 - 4 362 67
XM_021478250.1 XP_021333925.1 3 - 4 250 67
XM_021478251.1 XP_021333926.1 3 - 4 349 67
GenBank mRNA BC091794.1 - 4 - 5 399 67
BC093217.1 - 3 - 4 330 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005167224.4, XM_009304133.3, XM_021478249.1, XM_021478250.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
nucleic acid binding
RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0