This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03126295_m1
See other HSD17B12A GE Assays ›
Gene Symbol:
hsd17b12a
Gene Name:
hydroxysteroid (17-beta) dehydrogenase 12a
Gene Aliases:
KAR, fi06g11, wu:fi06g11, zgc:55589, zgc:77599
Chromosome Location:
Chr.25: 30196055 - 30230559 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03126295_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hsd17b12a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200881.1 NP_957175.1 1 - 2 298 102
GenBank mRNA AY551082.1 - 1 - 2 291 102
BC048053.1 - 1 - 2 240 102
BC063943.1 - 1 - 2 298 102
BC164345.1 - 1 - 2 215 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid biosynthetic process
steroid biosynthetic process
estrogen biosynthetic process
oxidation-reduction process
estradiol 17-beta-dehydrogenase activity
oxidoreductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0