This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03126088_g1
See other FZR1A GE Assays ›
Gene Symbol:
fzr1a
Gene Name:
fizzy/cell division cycle 20 related 1a
Gene Aliases:
fzr1, zgc:55450
Chromosome Location:
Chr.8: 384969 - 387702 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03126088_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fzr1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200253.1 NP_956547.1 8 - 9 949 73
GenBank mRNA BC048038.1 - 8 - 9 949 73
BC165357.1 - 7 - 8 757 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
anaphase-promoting complex-dependent catabolic process
cell division
positive regulation of ubiquitin protein ligase activity
positive regulation of anaphase-promoting complex-dependent catabolic process
anaphase-promoting complex binding
ubiquitin-protein transferase activator activity
ubiquitin ligase activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0