This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03118359_m1
See other MYO1CA GE Assays ›
Gene Symbol:
myo1ca
Gene Name:
myosin Ic, paralog a
Gene Aliases:
MMI-beta, MMIb, Myo1c, Myosin-Ic, si:ch211-69i14.4
Chromosome Location:
Chr.5: 37775924 - 37817146 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03118359_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: myo1ca
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001100031.1 NP_001093501.1 23 - 24 2428 62
XM_005165347.4 XP_005165404.1 23 - 24 2603 62
XM_005165349.4 XP_005165406.1 23 - 24 2663 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

actin binding motor protein

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
vesicle transport along actin filament
glomerulus development
microfilament motor activity
nucleotide binding
motor activity
actin binding
ATP binding
actin-dependent ATPase activity
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0