This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03118316_m1
See other SAP130A GE Assays ›
Gene Symbol:
sap130a
Gene Name:
Sin3A-associated protein a
Gene Aliases:
zgc:162722
Chromosome Location:
Chr.6: 633 - 13975 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03118316_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sap130a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098777.2 NP_001092247.2 15 - 16 2127 109
XM_005165896.1 XP_005165953.1 15 - 16 2136 109
XM_009302388.3 XP_009300663.1 15 - 16 2084 109
GenBank mRNA BC142598.1 - 15 - 16 2127 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein histone modifying enzyme

Gene Ontology Categories:

Function(s)

negative regulation of transcription from RNA polymerase II promoter
thigmotaxis
cardiac ventricle development
regulation of transcription, DNA-templated
larval locomotory behavior
floor plate formation

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0