This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03116979_m1
See other MFSD12A GE Assays ›
Gene Symbol:
mfsd12a
Gene Name:
major facilitator superfamily domain containing 12a
Gene Aliases:
fd45a11, si:dkey-218h11.4, wu:fd45a11, wz7872, zgc:158256
Chromosome Location:
Chr.2: 23080896 - 23099644 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03116979_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mfsd12a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044972.1 NP_001038437.1 4 - 5 964 127
XM_005171209.4 XP_005171266.1 4 - 5 1316 127

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate transport
pigment accumulation
organic substance transport
transporter activity
symporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0