This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03114817_g1
See other IMPDH1A GE Assays ›
Gene Symbol:
impdh1a
Gene Name:
IMP (inosine 5'-monophosphate) dehydrogenase 1a
Gene Aliases:
IMPD 1a, IMPDH 1a, zgc:91911
Chromosome Location:
Chr.18: 8804899 - 8842820 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03114817_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: impdh1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002177.1 NP_001002177.1 11 - 12 1348 73
XM_005159007.4 XP_005159064.1 11 - 12 1894 73
XM_005159008.4 XP_005159065.1 11 - 12 1896 73
XM_009293441.2 XP_009291716.1 11 - 12 1383 73
GenBank mRNA BC074090.1 - 11 - 12 1348 73
BC165226.1 - 10 - 11 1178 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

eye development
purine nucleotide biosynthetic process
GMP biosynthetic process
GTP biosynthetic process
developmental pigmentation
oxidation-reduction process
nucleotide binding
catalytic activity
IMP dehydrogenase activity
oxidoreductase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0