This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03112098_m1
See other MTCH2 GE Assays ›
Gene Symbol:
mtch2
Gene Name:
mitochondrial carrier homolog 2
Gene Aliases:
fi20c06, fj35g12, wu:fi20c06, wu:fj35g12
Chromosome Location:
Chr.7: 38667082 - 38678090 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03112098_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mtch2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131382.1 NP_571457.1 1 - 2 127 59
XM_021477393.1 XP_021333068.1 1 - 2 159 59
GenBank mRNA AF176010.1 - 1 - 2 127 59
BC055216.1 - 1 - 2 147 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s)

liver development
positive regulation of apoptotic process
fat cell differentiation
digestive tract development

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0