This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03110537_g1
See other PCSK5A GE Assays ›
Gene Symbol:
pcsk5a
Gene Name:
proprotein convertase subtilisin/kexin type 5a
Gene Aliases:
pc5.1, pcsk5.1, pcsk5/6a, si:dkey-267i17.1
Chromosome Location:
Chr.5: 127333 - 215542 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03110537_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pcsk5a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_021476634.1 XP_021332309.1 11 - 12 1676 86
GenBank mRNA DQ983232.1 - 11 - 12 1605 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
protein processing
peptide hormone processing
neuromast development
anterior lateral line development
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0