This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03110267_g1
See other DGKAA GE Assays ›
Gene Symbol:
dgkaa
Gene Name:
diacylglycerol kinase, alpha a
Gene Aliases:
dgka, zgc:136759
Chromosome Location:
Chr.23: 32239338 - 32304705 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03110267_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dgkaa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045315.1 NP_001038780.1 13 - 14 1318 67
XM_005162328.4 XP_005162385.1 14 - 15 1472 67
GenBank mRNA BC098607.1 - 13 - 14 1314 67
BC117645.1 - 13 - 14 1318 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
intracellular signal transduction
diacylglycerol metabolic process
glycerolipid metabolic process
lipid phosphorylation
nucleotide binding
NAD+ kinase activity
diacylglycerol kinase activity
calcium ion binding
ATP binding
kinase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0