This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03108360_m1
See other SLC38A5B GE Assays ›
Gene Symbol:
slc38a5b
Gene Name:
solute carrier family 38 member 5b
Gene Aliases:
slc38a3, wu:fc31c02, wu:fc48a10, zgc:92015
Chromosome Location:
Chr.8: 23580850 - 23612428 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03108360_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slc38a5b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002648.1 NP_001002648.1 15 - 16 1549 65
GenBank mRNA BC076529.1 - 15 - 16 1549 65
BC165220.1 - 14 - 15 1348 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
glutamine transport
glycine transport
serine transport
L-histidine transmembrane transport
amino acid transmembrane transporter activity
L-glutamine transmembrane transporter activity
glycine transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0