This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03107864_g1
See other TCIRG1B GE Assays ›
Gene Symbol:
tcirg1b tcirg1a
Gene Name:
T cell immune regulator 1, ATPase H+ transporting V0 subunit a3b
T cell immune regulator 1, ATPase H+ transporting V0 subunit a3a
Gene Aliases:
tcirg1, wu:fb44e06, wu:fi46h07, zgc:55891
si:dkey-9i23.9, tcirg1
Chromosome Location:
Chr.14: 16777713 - 16810439 on Build GRCz11
Chromosome Location:
Chr.1: 44872299 - 44901111 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
-
Assay ID Dr03107864_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tcirg1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213069.1 NP_998234.1 12 - 13 1583 97
GenBank mRNA BC045484.1 - 12 - 13 1583 97
BC165671.1 - 11 - 12 1496 97
Gene Symbol: tcirg1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA CN014012.1 - 1 - 2 208 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ion transport
vacuolar acidification
hydrogen ion transmembrane transport
hydrogen ion transmembrane transporter activity
proton-transporting ATPase activity, rotational mechanism
ATPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0