This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03105756_g1
See other DNMT3BB.2 GE Assays ›
Gene Symbol:
dnmt3bb.2
Gene Name:
DNA (cytosine-5-)-methyltransferase 3 beta, duplicate b.2
Gene Aliases:
Zmt3, dnmt3, wu:fa11d06, wu:fb73b09
Chromosome Location:
Chr.23: 16983363 - 17004783 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03105756_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dnmt3bb.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131386.1 NP_571461.1 5 - 6 2133 95
GenBank mRNA AB196914.1 - 5 - 6 2133 95
AF135438.1 - 1 - 2 119 95
BC162467.1 - 5 - 6 2022 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

DNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

DNA methylation
retina layer formation
neurogenesis
exocrine pancreas development
methylation
DNA methylation on cytosine
eye photoreceptor cell development
regulation of histone H3-K9 methylation
pharyngeal system development
retina morphogenesis in camera-type eye
methyltransferase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0