This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03103662_m1
See other SLC26A3.2 GE Assays ›
Gene Symbol:
slc26a3.2
Gene Name:
solute carrier family 26 member 3, tandem duplicate 2
Gene Aliases:
si:dkey-31f5.1, zgc:136981
Chromosome Location:
Chr.4: 18799197 - 18823032 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03103662_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slc26a3.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040175.1 NP_001035265.1 10 - 11 1315 72
XM_005164755.4 XP_005164812.1 10 - 11 1438 72
GenBank mRNA BC115355.1 - 10 - 11 1315 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

chloride transport
sulfate transport
bicarbonate transport
transmembrane transport
secondary active sulfate transmembrane transporter activity
bicarbonate transmembrane transporter activity
chloride transmembrane transporter activity
sulfate transmembrane transporter activity
antiporter activity
anion:anion antiporter activity
oxalate transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0