This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03091249_g1
See other INPPL1A GE Assays ›
Gene Symbol:
inppl1a
Gene Name:
inositol polyphosphate phosphatase-like 1a
Gene Aliases:
INPPL1-A, SHIP-2A, fc58a04, ship2a, wu:fc58a04, zgc:198309
Chromosome Location:
Chr.15: 47454443 - 47498275 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03091249_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: inppl1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039804.2 NP_001034893.1 15 - 16 2376 110
GenBank mRNA BC171582.1 - 16 - 17 2296 110
DQ272661.2 - 15 - 16 2376 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

immune system process
cell adhesion
BMP signaling pathway
negative regulation of fibroblast growth factor receptor signaling pathway
phosphatidylinositol dephosphorylation
negative regulation of neutrophil chemotaxis
actin binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0