This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03089444_m1
See other RGS7A GE Assays ›
Gene Symbol:
rgs7a
Gene Name:
regulator of G protein signaling 7a
Gene Aliases:
rgs7, zgc:92793
Chromosome Location:
Chr.17: 19626465 - 19727116 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03089444_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rgs7a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002541.1 NP_001002541.1 7 - 8 643 63
XM_009292967.3 XP_009291242.1 8 - 9 799 63
XM_017351624.2 XP_017207113.1 8 - 9 797 63
XM_021467140.1 XP_021322815.1 8 - 9 800 63
XM_021467141.1 XP_021322816.1 7 - 8 585 63
GenBank mRNA BC076267.1 - 7 - 8 643 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s)

G-protein coupled receptor signaling pathway
intracellular signal transduction

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0