This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03088098_g1
See other PITPNAA GE Assays ›
Gene Symbol:
pitpnaa
Gene Name:
phosphatidylinositol transfer protein, alpha a
Gene Aliases:
Pitpn, pitpna, zgc:55569
Chromosome Location:
Chr.15: 28410625 - 28425011 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03088098_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pitpnaa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200935.1 NP_957229.1 9 - 10 742 76
XM_005157645.4 XP_005157702.1 9 - 10 826 76
GenBank mRNA BC044192.1 - 9 - 10 742 76
BC163953.1 - 9 - 10 677 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

axonogenesis
phospholipid transport
chordate embryonic development
phospholipid transporter activity
phosphatidylcholine transporter activity
phosphatidylinositol transporter activity
phosphatidylcholine binding
phosphatidylinositol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0