This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03084275_m1
See other FGF20A GE Assays ›
Gene Symbol:
fgf20a
Gene Name:
fibroblast growth factor 20a
Gene Aliases:
dob, wu:fb05c08
Chromosome Location:
Chr.1: 36692 - 38755 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03084275_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fgf20a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037103.2 NP_001032180.1 1 - 2 364 60
GenBank mRNA BC162861.1 - 1 - 2 364 60
BC162886.1 - 1 - 2 364 60
DQ195158.1 - 1 - 2 271 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
organ morphogenesis
positive regulation of gene expression
cell differentiation
regulation of cell migration
fin regeneration
regulation of bone remodeling
anatomical structure homeostasis
fibroblast growth factor receptor binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0