This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03075823_m1
See other POLL GE Assays ›
Gene Symbol:
poll
Gene Name:
polymerase (DNA directed), lambda
Gene Aliases:
wu:fe49a04, zgc:66320, zgc:77901
Chromosome Location:
Chr.1: 31657880 - 31672343 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03075823_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: poll
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213243.1 NP_998408.1 5 - 6 921 68
GenBank mRNA BC055597.1 - 5 - 6 959 68
BC065469.1 - 5 - 6 921 68
BC165409.1 - 4 - 5 908 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed DNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
double-strand break repair via nonhomologous end joining
DNA binding
DNA-directed DNA polymerase activity
transferase activity
nucleotidyltransferase activity
DNA polymerase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0