This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm03419548_g1
See other SU(F) GE Assays ›
Gene Symbol:
su(f)
Gene Name:
suppressor of forked
Gene Aliases:
19, 28, 29, 3-DES, 3DES, CG17170, CstF-77, CstF77, Cstf77, Dmel\CG17170, Dmel_CG17170, EC235, R-9-18, R9-18, SU-f, Su(f), l(1)20Eb, l(1)3DES, l(1)EC235, l(1)G0393, l(1)R-9-18, l(1)X1, l(1)X3, l(1)su(f), l3DES, su-f, su[+]-f
Chromosome Location:
Chr.X: 23096771 - 23101431 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0003559
Assay ID Dm03419548_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: su(f)
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001015241.4 NP_001015241.3 3 - 4 916 87
NM_001111009.4 NP_001104479.1 3 - 4 916 87
NM_001111010.3 NP_001104480.2 4 - 5 1010 87
GenBank mRNA BT021384.1 - 3 - 4 933 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Drosophila melanogaster

Panther Classification:

Molecular Function -

mRNA polyadenylation factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA 3'-end processing
positive regulation of malate dehydrogenase (decarboxylating) (NADP+) activity
mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0