This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02541654_s1
See other SDS22 GE Assays ›
Gene Symbol:
sds22
Gene Name:
CG5851-PA
Gene Aliases:
CG5851, CT18355, Dmel\CG5851, Dmel_CG5851, Sds22
Chromosome Location:
Chr.3R: 17391594 - 17393020 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0028992
Assay ID Dm02541654_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sds22
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_142362.4 NP_650619.1 2 1227 68
GenBank mRNA AY070854.1 - 2 1155 68
AY118751.1 - 1 1052 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase modulator

Gene Ontology Categories:

Function(s)

negative regulation of cell proliferation
establishment or maintenance of polarity of follicular epithelium
establishment or maintenance of polarity of larval imaginal disc epithelium

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0