This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02373718_s1
See other HTL GE Assays ›
Gene Symbol:
htl
Gene Name:
heartless
Gene Aliases:
CG7223, CT22273, CT39172, DFGF-R1, DFGF-R2, DFR-1, DFR1, DFR1/DFGF-R2, DPR3, DTRK(FR1), Dfr-1, Dfr1, DmHD-38, Dmel\CG7223, Dmel_CG7223, Dtk1, EMS2, FGF-R2, FGFR, FGFR2, FR1, Fr1, HD-38, HTL/FGFR1, Htl, Tk1, dtk1, i100, i150, i79, j372
Chromosome Location:
Chr.3R: 18045216 - 18052984 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0010389
Assay ID Dm02373718_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: htl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_079670.3 NP_524394.2 2 1863 59
NM_169784.2 NP_732286.1 2 1636 59
NM_169785.2 NP_732287.1 2 1517 59
GenBank mRNA AY051812.1 - 2 1835 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

mesodermal cell fate commitment
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
multicellular organism development
pole cell migration
central nervous system development
ventral cord development
salivary gland development
endodermal cell fate determination
mesodermal cell fate determination
gonadal mesoderm development
heart development
mesoderm migration involved in gastrulation
visceral muscle development
larval visceral muscle development
somatic muscle development
mesodermal cell migration
positive regulation of cell proliferation
glial cell migration
germ cell migration
regulation of cell shape
fibroblast growth factor receptor signaling pathway
glial cell differentiation
cardioblast differentiation
glial cell development
positive regulation of kinase activity
lymph gland plasmatocyte differentiation
lymph gland crystal cell differentiation
glial cell growth
positive regulation of innate immune response
positive regulation of synaptic growth at neuromuscular junction
lymph gland development
myoblast fate specification
muscle fiber development
defense response to Gram-negative bacterium
pericardial nephrocyte differentiation
positive regulation of glial cell migration
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
protein binding
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0