This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02371390_s1
See other EIF4E7 GE Assays ›
Gene Symbol:
eIF4E7
Gene Name:
eukaryotic translation initiation factor 4E7
Gene Aliases:
CG11392, CG11393, CG32859, CT31794, Dmel\CG32859, Dmel_CG32859, EG:BACR42I17.1, eIF4E, eIF4E-7
Chromosome Location:
Chr.X: 1158599 - 1162389 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0040368
Assay ID Dm02371390_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eIF4E7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_166870.2 NP_726718.1 2 1558 95
GenBank mRNA BT014927.1 - 1527 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster Inventoried

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
RNA cap binding
RNA 7-methylguanosine cap binding
translation initiation factor activity
protein binding
eukaryotic initiation factor 4G binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0