This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02332497_g1
See other DPY GE Assays ›
Gene Symbol:
dpy
Gene Name:
dumpy
Gene Aliases:
BcDNA:SD02173, CG15637, CG31724, CG31725, CG31726, CG31727, CG31964, CG31965, CG31966, CG31967, CG31968, CG33000, CG33086, CG33196, CR33195, CT35799, DP, DPY, Dmel\CG33196, Dmel_CG33196, Dp, Dpy, Mur24F, Q8IQ18_DROME, SP460, dp, l(2)24Fd, olv[D]
Chromosome Location:
Chr.2L: 4477462 - 4614300 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0053196
Assay ID Dm02332497_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dpy
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001258946.1 NP_001245875.1 40 - 41 40398 150
NM_001258950.1 NP_001245879.1 33 - 34 19221 150
NM_001273101.1 NP_001260030.1 75 - 76 61703 150
NM_001273102.1 NP_001260031.1 59 - 60 55614 150
NM_001273103.1 NP_001260032.1 75 - 76 62331 150
NM_001273104.1 NP_001260033.1 74 - 75 61974 150
NM_001273105.1 NP_001260034.1 71 - 72 49407 150
NM_001273106.1 NP_001260035.1 69 - 70 60384 150
NM_001273107.1 NP_001260036.1 64 - 65 58455 150
NM_001273108.1 NP_001260037.1 73 - 74 54696 150
NM_001273109.1 NP_001260038.1 57 - 58 47516 150
NM_001273111.1 NP_001260040.1 69 - 70 47759 150
NM_001273112.1 NP_001260041.1 64 - 65 41478 150
NM_001273113.1 NP_001260042.1 60 - 61 52164 150

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
open tracheal system development
apposition of dorsal and ventral imaginal disc-derived wing surfaces
chitin-based embryonic cuticle biosynthetic process
chitin-based cuticle attachment to epithelium
regulation of imaginal disc-derived wing size
transcription factor activity, sequence-specific DNA binding
extracellular matrix structural constituent
calcium ion binding
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0