This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02152015_m1
See other HEPH GE Assays ›
Gene Symbol:
heph
Gene Name:
hephaestus
Gene Aliases:
CG2094, CG2290, CG31000, DmPTB, Dmel\CG31000, Dmel_CG31000, HEPH, Heph, PTB, Ptb, dPTB, dmPTB, ema, hnRNP I, l(3)03429, l(3)j11B9, ptb
Chromosome Location:
Chr.3R: 31846987 - 32015520 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0011224
Assay ID Dm02152015_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: heph
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001104522.3 NP_001097992.1 10 - 11 1713 82
NM_001104523.3 NP_001097993.1 12 - 13 2533 82
NM_001104524.3 NP_001097994.1 10 - 11 1978 82
NM_001170316.2 NP_001163787.1 12 - 13 2103 82
NM_001170317.2 NP_001163788.1 10 - 11 1701 82
NM_001170318.2 NP_001163789.1 7 - 8 1500 82
NM_001170319.1 NP_001163790.1 11 - 12 2141 82
NM_001170320.2 NP_001163791.1 8 - 9 2360 82
NM_001170321.2 NP_001163792.1 9 - 10 1802 82
NM_001170322.2 NP_001163793.1 9 - 10 2382 82
NM_001170323.2 NP_001163794.1 11 - 12 2237 82
NM_001260469.2 NP_001247398.1 12 - 13 2296 82
NM_001260470.1 NP_001247399.1 13 - 14 2514 82
NM_001300701.1 NP_001287630.1 10 - 11 2592 82
NM_079964.3 NP_524703.1 8 - 9 1624 82
NM_170581.2 NP_733460.1 10 - 11 1514 82
NM_170582.4 NP_733461.2 10 - 11 1978 82
NM_176596.3 NP_788773.1 11 - 12 1560 82
NM_176597.3 NP_788774.1 10 - 11 2466 82
NM_176598.2 NP_788775.1 10 - 11 2072 82
NM_176599.2 NP_788776.2 10 - 11 1631 82
NM_176600.3 NP_788777.1 9 - 10 1671 82
NM_176601.3 NP_788778.1 9 - 10 1729 82
NM_176602.3 NP_788779.1 9 - 10 1491 82
NM_176603.3 NP_788780.1 12 - 13 2103 82
GenBank mRNA AF211191.2 - 8 - 9 1646 82
AF436844.1 - 12 - 13 2149 82
AF455053.1 - 10 - 11 1514 82
AY052367.1 - 12 - 13 2094 82
BT030984.1 - 12 - 13 2086 82
BT032832.1 - 10 - 11 1688 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
spermatid development
sperm individualization
negative regulation of oskar mRNA translation
oocyte microtubule cytoskeleton polarization
imaginal disc-derived wing vein morphogenesis
imaginal disc-derived wing margin morphogenesis
translation repressor activity, nucleic acid binding
mRNA binding
mRNA 3'-UTR binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0