This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02151330_m1
See other CRB GE Assays ›
Gene Symbol:
crb
Gene Name:
crumbs
Gene Aliases:
0509/20, 1384/04, CG6383, CRB, CT19912, Crb, Crbs, Crumbs, DmCrb, Dmel\CG6383, Dmel_CG6383, crumb, far, l(3)07207, l(3)S050920, l(3)S058104, l(3)j1B5
Chromosome Location:
Chr.3R: 24295078 - 24314541 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0259685
Assay ID Dm02151330_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: crb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_079756.3 NP_524480.2 3 - 4 2121 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

compound eye morphogenesis
apical constriction
establishment or maintenance of cell polarity
dorsal closure
nervous system development
open tracheal system development
salivary gland morphogenesis
Malpighian tubule morphogenesis
positive regulation of cell proliferation
establishment or maintenance of polarity of embryonic epithelium
establishment or maintenance of polarity of follicular epithelium
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of protein localization
regulation of intracellular protein transport
adherens junction organization
liquid clearance, open tracheal system
establishment or maintenance of apical/basal cell polarity
maintenance of apical/basal cell polarity
tube morphogenesis
positive regulation of hippo signaling
compound eye photoreceptor development
rhabdomere development
apical protein localization
zonula adherens assembly
zonula adherens maintenance
photoreceptor cell maintenance
regulation of imaginal disc growth
negative regulation of Notch signaling pathway
negative regulation of organ growth
dorsal closure, amnioserosa morphology change
amnioserosa maintenance
wing disc dorsal/ventral pattern formation
protein stabilization
centrosome localization
trachea development
membrane organization
cell morphogenesis involved in Malpighian tubule morphogenesis
rhabdomere morphogenesis
nuclear chromosome segregation
assembly of apicomedial cortex actomyosin
protein kinase C binding
Notch binding
calcium ion binding
spectrin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0