This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02150661_g1
See other TWIN GE Assays ›
Gene Symbol:
twin
Gene Name:
CG31137-PA
Gene Aliases:
BEST:GH20274, CCR4, CCR4/Twin, CG17741, CG31137, CG5534, CT39331, Ccr4, Dmel\CG31137, Dmel_CG31137, Twin, ccr4, twn
Chromosome Location:
Chr.3R: 24196893 - 24221965 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0011725
Assay ID Dm02150661_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: twin
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_170128.2 NP_732964.1 7 - 8 962 80
NM_170129.3 NP_732965.1 7 - 8 1078 80
NM_170130.3 NP_732966.1 7 - 8 1226 80
NM_170131.3 NP_732967.1 6 - 7 1194 80
NM_170132.3 NP_732968.1 3 - 4 1326 80
NM_176555.2 NP_788732.1 7 - 8 1066 80
GenBank mRNA AY069458.1 - 7 - 8 1054 80
BT001344.1 - 6 - 7 1092 80
BT010039.1 - 7 - 8 962 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Drosophila melanogaster

Panther Classification:

Molecular Function -

mRNA polyadenylation factor

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA poly(A) tail shortening
mRNA catabolic process
negative regulation of translation
female germ-line stem cell asymmetric division
oogenesis
3'-5'-exoribonuclease activity
poly(A)-specific ribonuclease activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0