This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm02146986_g1
See other LK6 GE Assays ›
Gene Symbol:
Lk6
Gene Name:
Lk6 kinase
Gene Aliases:
CG17342, CG6929, Dmel\CG17342, Dmel_CG17342, LK6, MESR8, MNK/LK6, anon-WO0172774.84, lk6
Chromosome Location:
Chr.3R: 11750931 - 11764457 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0017581
Assay ID Dm02146986_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lk6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_143729.3 NP_651986.2 3 - 4 1664 63
NM_169423.3 NP_731624.1 3 - 4 1216 63
GenBank mRNA BT050542.1 - 3 - 4 989 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
microtubule-based process
peptidyl-serine phosphorylation
positive regulation of peptidyl-serine phosphorylation
intracellular signal transduction
regulation of growth
protein autophosphorylation
protein kinase activity
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
calmodulin binding
ATP binding
calcium-dependent protein serine/threonine kinase activity
kinase activity
mitogen-activated protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0