This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01846494_g1
See other GYC32E GE Assays ›
Gene Symbol:
Gyc32E
Gene Name:
Guanylyl cyclase at 32E
Gene Aliases:
CG33114, CG6275, Cyg1, DMRGC, Dmel\CG33114, Dmel_CG33114, GC, GYC 32E, Gyc-r
Chromosome Location:
Chr.2L: 11138759 - 11149101 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0010197
Assay ID Dm01846494_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gyc32E
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001103678.2 NP_001097148.1 19 - 20 2860 98
NM_001298905.1 NP_001285834.1 18 - 19 2752 98
NM_176013.2 NP_788027.1 18 - 19 2776 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Drosophila melanogaster

Panther Classification:

Molecular Function -

guanylate cyclase

Gene Ontology Categories:

Function(s) Process(es)

cGMP biosynthetic process
protein phosphorylation
signal transduction
receptor guanylyl cyclase signaling pathway
cGMP-mediated signaling
peptide receptor activity
guanylate cyclase activity
protein kinase activity
ATP binding
signaling receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0