This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01842578_g1
See other SR-CI GE Assays ›
Gene Symbol:
Sr-CI
Gene Name:
Scavenger receptor class C, type I
Gene Aliases:
CG4099, CT9876, D-SR-CI, DSR-CI, Dmel\CG4099, Dmel_CG4099, SR-C1, SR-CI, Sr-C1, Sr-CL, Sr-Cl, dSR-C1, dSR-CI, dSRC-I, dScR-I
Chromosome Location:
Chr.2L: 4121702 - 4124153 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0014033
Assay ID Dm01842578_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sr-CI
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001298670.1 NP_001285599.1 2 - 3 815 81
NM_057754.3 NP_477102.1 2 - 3 815 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
phagocytosis
immune response
response to bacterium
defense response to Gram-negative bacterium
scavenger receptor activity
polysaccharide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0