This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01842556_m1
See other CNN GE Assays ›
Gene Symbol:
cnn
Gene Name:
centrosomin
Gene Aliases:
CG 4832, CG18370, CG4832, CNN, Cnn, Cnn1, Dmel\CG4832, Dmel_CG4832, cen, mat(2)syn-C, mat(2)synHK21, mfs, obv
Chromosome Location:
Chr.2R: 13439314 - 13450475 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0013765
Assay ID Dm01842556_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cnn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001259363.2 NP_001246292.1 1 - 2 304 66
NM_001259364.2 NP_001246293.1 1 - 2 304 66
NM_057736.6 NP_477084.1 1 - 2 304 66
GenBank mRNA BT010053.1 - 1 - 2 274 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

mitotic spindle organization
centrosome cycle
centriole replication
pole cell formation
central nervous system development
peripheral nervous system development
midgut development
asymmetric cell division
photoreceptor cell morphogenesis
regulation of centriole-centriole cohesion
embryonic cleavage
asymmetric neuroblast division
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0