This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01834384_g1
See other CG7028 GE Assays ›
Gene Symbol:
CG7028
Gene Name:
CG7028-PA
Gene Aliases:
BcDNA:GH04978, Dmel\CG7028, Dmel_CG7028, PRP4, PRP4K, prp4
Chromosome Location:
Chr.3L: 207184 - 211457 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0027587
Assay ID Dm01834384_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CG7028
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001274257.1 NP_001261186.1 5 - 6 2037 109
NM_001299955.1 NP_001286884.1 5 - 6 2040 109
NM_001299956.1 NP_001286885.1 5 - 6 2022 109
NM_001299957.1 NP_001286886.1 5 - 6 2025 109
NM_138166.4 NP_612010.1 5 - 6 2025 109
GenBank mRNA AF145626.1 - 5 - 6 1998 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
protein phosphorylation
positive regulation of hippo signaling
protein kinase activity
protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0