This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01825650_m1
See other SYD GE Assays ›
Gene Symbol:
syd
Gene Name:
sunday driver
Gene Aliases:
CG8110, Dmel\CG8110, Dmel_CG8110, JIP-3, Syd, dsyd-1, jip3
Chromosome Location:
Chr.3L: 7916650 - 7936120 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0024187
Assay ID Dm01825650_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: syd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001169902.2 NP_001163373.1 1 - 2 745 68
NM_079913.3 NP_524652.1 1 - 2 745 68
GenBank mRNA AF262045.1 - 1 - 2 499 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

activation of JUN kinase activity
axo-dendritic transport
vesicle-mediated transport
regulation of JNK cascade
MAP-kinase scaffold activity
JUN kinase binding
kinesin binding
receptor signaling complex scaffold activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0