This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01822360_g1
See other RPL14 GE Assays ›
Gene Symbol:
RpL14
Gene Name:
Ribosomal protein L14
Gene Aliases:
CG6253, Dmel\CG6253, Dmel_CG6253, L14, M(3), M(3)66, M(3)66D, M66D, RPL14, Rp L14, anon-EST:Posey193U, anon-EST:Posey6, rpl14
Chromosome Location:
Chr.3L: 8601006 - 8602392 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0017579
Assay ID Dm01822360_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RpL14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_079251.4 NP_523975.1 3 - 4 331 57
GenBank mRNA KX531263.1 - 2 - 3 302 57
Y09766.1 - 313 57
Y10017.1 - 307 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

cytoplasmic translation
translation
ribosomal large subunit biogenesis
negative regulation of neuron apoptotic process
RNA binding
structural constituent of ribosome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0