This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01821178_g1
See other TROL GE Assays ›
Gene Symbol:
trol
Gene Name:
terribly reduced optic lobes
Gene Aliases:
BcDNA:GM02481, CG12497, CG33675, CG33950, CG7981, CT23996, Dmel\CG33950, Dmel_CG33950, EG:BACR25B3.1, EG:BACR25B3.10, EG:BACR25B3.11, EG:BACR25B3.2, GC7891, MRE7, Pcan, Pcn, TROL, Trol, Trol-A, Trol-B, Troll, ZW-1, anon-WO0153538.72, l(1)3Ac, l(1)9-96, l(1)G0019, l(1)G0021, l(1)G0023, l(1)G0181, l(1)G0211, l(1)G0271, l(1)G0374, l(1)G0412, l(1)VA51, l(1)trol, l(1)zw1, l(1)zwl, pcan, troll, zw-1, zw1
Chromosome Location:
Chr.X: 2470460 - 2545427 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0284408
Assay ID Dm01821178_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: trol
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031862.4 NP_001027033.3 20 - 21 6992 65
NM_001031863.5 NP_001027034.2 27 - 28 7663 65
NM_001031864.4 NP_001027035.3 19 - 20 6632 65
NM_001031865.4 NP_001027036.3 20 - 21 6992 65
NM_001031866.4 NP_001027037.3 22 - 23 7421 65
NM_001031867.4 NP_001027038.3 29 - 30 8324 65
NM_001103399.4 NP_001096869.3 19 - 20 6938 65
NM_001169177.4 NP_001162648.3 28 - 29 8204 65
NM_001258562.3 NP_001245491.2 29 - 30 8324 65
NM_001258563.3 NP_001245492.2 16 - 17 6155 65
NM_001258564.3 NP_001245493.2 28 - 29 8273 65
NM_001258565.3 NP_001245494.2 18 - 19 6470 65
NM_001258566.4 NP_001245495.2 27 - 28 7506 65
NM_001258567.3 NP_001245496.2 17 - 18 6245 65
NM_001258568.3 NP_001245497.2 19 - 20 6872 65
NM_001258569.3 NP_001245498.2 17 - 18 6389 65
NM_001258570.3 NP_001245499.2 25 - 26 7868 65
NM_001258571.3 NP_001245500.2 22 - 23 7421 65
NM_001297893.2 NP_001284822.1 17 - 18 6386 65
NM_001297894.2 NP_001284823.1 19 - 20 6890 65
NM_001297895.2 NP_001284824.1 17 - 18 6236 65
NM_001297896.2 NP_001284825.1 23 - 24 7580 65
NM_001382068.1 NP_001368970.1 17 - 18 6245 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Gene Ontology Categories:

Function(s)

positive regulation of neuroblast proliferation
defasciculation of motor neuron axon
motor neuron axon guidance
organ morphogenesis
tissue development
negative regulation of fibroblast growth factor receptor signaling pathway
maintenance of epithelial cell apical/basal polarity
positive regulation of fibroblast growth factor receptor signaling pathway
positive regulation of smoothened signaling pathway
response to anesthetic
positive regulation of semaphorin-plexin signaling pathway

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0