This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01821036_g1
See other ADAR GE Assays ›
Gene Symbol:
Adar
Gene Name:
Adenosine deaminase acting on RNA
Gene Aliases:
ADAR, ADAR1, CG12598, Dmel\CG12598, Dmel_CG12598, EG:BACN35H14.1, adar, adr, cg12598, dADAR, dAdar, dadar, hypnos-2
Chromosome Location:
Chr.X: 1773725 - 1788065 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0026086
Assay ID Dm01821036_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adar
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038732.3 NP_001033821.1 4 - 5 774 61
NM_001258540.1 NP_001245469.1 4 - 5 853 61
NM_001258541.2 NP_001245470.1 3 - 4 710 61
NM_001258542.2 NP_001245471.1 4 - 5 823 61
NM_001258543.2 NP_001245472.2 4 - 5 774 61
NM_001258544.2 NP_001245473.1 4 - 5 925 61
NM_001258545.2 NP_001245474.1 3 - 4 707 61
NM_001258546.2 NP_001245475.1 4 - 5 774 61
NM_001258547.2 NP_001245476.1 5 - 6 965 61
NM_001258548.2 NP_001245477.1 4 - 5 1016 61
NM_001297862.1 NP_001284791.1 4 - 5 774 61
NM_001297863.1 NP_001284792.1 3 - 4 734 61
NM_001382020.1 NP_001368969.1 3 - 4 441 61
NM_130584.4 NP_569940.2 4 - 5 774 61
NM_166903.3 NP_726761.2 3 - 4 707 61
GenBank mRNA AF343579.1 - 4 - 5 755 61
AY118653.1 - 4 - 5 749 61
BT099919.1 - 4 - 5 719 61
GQ168940.1 - 4 - 5 736 61
GQ168941.1 - 4 - 5 725 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
adenosine to inosine editing
RNA processing
response to oxidative stress
locomotory behavior
male courtship behavior
adult locomotory behavior
response to heat
RNA modification
mRNA modification
adult behavior
regulation of membrane potential
regulation of circadian rhythm
locomotor rhythm
regulation of gene silencing by miRNA
double-stranded RNA binding
double-stranded RNA adenosine deaminase activity
tRNA-specific adenosine deaminase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0