This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01816737_g1
See other HPPY GE Assays ›
Gene Symbol:
hppy
Gene Name:
happyhour
Gene Aliases:
CG7097, CG7097/hppy, Dmel\CG7097, Dmel_CG7097, EP-704, GLK/KHS1, Hppy, MAP4K3, anon-WO0153538.68, l(2)SH1261, l(2)SH2 1261
Chromosome Location:
Chr.2R: 19178738 - 19227193 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0263395
Assay ID Dm01816737_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hppy
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001299675.1 NP_001286604.1 14 - 15 3052 74
NM_001299676.1 NP_001286605.1 12 - 13 3763 74
NM_001299677.1 NP_001286606.1 12 - 13 2950 74
NM_137555.3 NP_611399.2 13 - 14 2989 74
NM_166335.3 NP_725863.1 13 - 14 3802 74
GenBank mRNA BT150295.1 - 13 - 14 2973 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
apoptotic process
Golgi organization
JNK cascade
regulation of cell size
positive regulation of TOR signaling
positive regulation of hippo signaling
regulation of growth rate
negative regulation of epidermal growth factor receptor signaling pathway
positive regulation of apoptotic process
positive regulation of cell size
positive regulation of JNK cascade
behavioral response to ethanol
triglyceride homeostasis
positive regulation of TORC1 signaling
protein kinase activity
protein serine/threonine kinase activity
ATP binding
MAP kinase kinase kinase kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0