This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01816679_m1
See other 5-HT1A GE Assays ›
Gene Symbol:
5-HT1A
Gene Name:
5-hydroxytryptamine (serotonin) receptor 1A
Gene Aliases:
5-HT-dro2A, 5-HT1ADro, 5-HT1a, 5-HT[1A]Dro, 5-HT[[1ADro]], 5-HT[[1A]], 5-HT[[1A]]DRO, 5HT-1A, 5HT-R2A, 5HT-dro2A, 5HT-drp[[2A]], 5HT1A, 5HT1a, 5HT[[1ADro]], 5htdro2a, CG16720, DRO2A, Dm5-HT1A, Dm5-HT[[1A]], Dm5HTdro2A, Dmel\CG16720, Dmel_CG16720, HT1A, d5-HT1A
Chromosome Location:
Chr.2R: 19067155 - 19121958 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0004168
Assay ID Dm01816679_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 5-HT1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001369983.1 NP_001356890.1 4 - 5 1916 77
NM_166322.2 NP_725849.1 4 - 5 1916 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

inter-male aggressive behavior
G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
adenylate cyclase-inhibiting serotonin receptor signaling pathway
phospholipase C-activating serotonin receptor signaling pathway
serotonin receptor signaling pathway
chemical synaptic transmission
anesthesia-resistant memory
male courtship behavior
regulation of circadian sleep/wake cycle, sleep
Gi/o-coupled serotonin receptor activity
G-protein coupled receptor activity
G-protein coupled serotonin receptor activity
G-protein coupled amine receptor activity
neurotransmitter receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0