This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01816284_g1
See other DARK GE Assays ›
Gene Symbol:
Dark
Gene Name:
Death-associated APAF1-related killer
Gene Aliases:
APAF-1, APAF1, ARK, Apaf, Apaf-1, Apaf1, Ark, CG6829, D-Apaf-1, DARK, Dapaf-1, Dapaf-1/HAC-1, Dark/Apaf-I, Dark/Dapaf-1/HAC1, Dark/Hac-1/dApaf-1, Dark/Hac-1/dApaf1, Dmel\CG6829, Dmel_CG6829, Hac-1, Hac-1/Dark, Hac1, T1, anon-53Fa, apaf-1, apaf1, arc, ark, dAPAF-1, dApaf-1, dApaf-1/DARK/HAC-1, dApaf1, dArk, dapaf, dapaf-1, dapaf-1L, dapaf-1S, dark, dark/dapaf-1/hac-1, dark/hac-1/dapaf-1, hac-1, hac1, l(2)SH0173
Chromosome Location:
Chr.2R: 17020027 - 17026703 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0263864
Assay ID Dm01816284_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dark
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_166207.3 NP_725637.1 6 - 7 1819 66
GenBank mRNA AB027531.1 - 6 - 7 1781 66
AF162659.1 - 6 - 7 1670 66
AF196306.2 - 5 - 6 1604 66
BT150397.1 - 6 - 7 1802 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of humoral immune response
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
sperm individualization
activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
response to gamma radiation
programmed cell death
central nervous system formation
chaeta development
S-adenosylmethionine cycle
positive regulation of glial cell apoptotic process
melanization defense response
salivary gland histolysis
salivary gland cell autophagic cell death
response to starvation
positive regulation of apoptotic process
positive regulation of compound eye retinal cell programmed cell death
dendrite morphogenesis
neuron cellular homeostasis
triglyceride homeostasis
sarcosine catabolic process
ATP binding
cysteine-type endopeptidase activator activity involved in apoptotic process
ADP binding
CARD domain binding
cysteine-type endopeptidase activity involved in apoptotic signaling pathway

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0