This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01809596_g1
See other OSTGAMMA GE Assays ›
Gene Symbol:
Ostgamma
Gene Name:
Oligosaccharide transferase gamma subunit
Gene Aliases:
CG7830, Dmel\CG7830, Dmel_CG7830, MagT1, OST, dMagT1
Chromosome Location:
Chr.2L: 8306696 - 8308192 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0032015
Assay ID Dm01809596_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ostgamma
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_135360.4 NP_609204.2 1 - 2 209 71
GenBank mRNA AY075331.1 - 1 - 2 191 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s)

cell migration
protein N-linked glycosylation via asparagine
encapsulation of foreign target
magnesium ion transmembrane transport

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0