This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01809081_m1
See other TTLL3A GE Assays ›
Gene Symbol:
TTLL3A
Gene Name:
Tubulin tyrosine ligase-like 3A
Gene Aliases:
CG11323, DmCG11323, Dmel\CG11323, Dmel_CG11323, TTLL3, dmTTLL3A
Chromosome Location:
Chr.2L: 6736364 - 6740188 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0031854
Assay ID Dm01809081_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TTLL3A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_135225.2 NP_609069.1 1 - 2 1394 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

protein polyglycylation
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0