This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01809080_g1
See other TTLL3B GE Assays ›
Gene Symbol:
TTLL3B
Gene Name:
Tubulin tyrosine ligase-like 3B
Gene Aliases:
CG11201, DmCG11201, Dmel\CG11201, Dmel_CG11201, TTLL3, dmTTLL3B
Chromosome Location:
Chr.2L: 6732866 - 6736149 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0031853
Assay ID Dm01809080_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TTLL3B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_135224.3 NP_609068.2 1 - 2 1714 84
GenBank mRNA BT021443.1 - 1714 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster

Gene Ontology Categories:

Function(s) Process(es)

spermatogenesis
sperm individualization
protein polyglycylation
axoneme assembly
cilium morphogenesis
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0