This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01808381_g1
See other APOLTP GE Assays ›
Gene Symbol:
Apoltp
Gene Name:
Apolipoprotein lipid transfer particle
Gene Aliases:
ApoLTPI, ApoLTPII, CG15828, CT39214, Dmel\CG15828, Dmel_CG15828, LTP, apoLTP, apoltp
Chromosome Location:
Chr.2L: 9634179 - 9656026 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0032136
Assay ID Dm01808381_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apoltp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_205948.3 NP_995670.2 14 - 15 11447 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Drosophila melanogaster

Panther Classification:

Molecular Function -

storage protein

Gene Ontology Categories:

Function(s) Process(es)

sterol transport
positive regulation of lipid transport
acylglycerol transport
lipid transporter activity
lipoprotein particle receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0