This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dm01804543_g1
See other GMD GE Assays ›
Gene Symbol:
Gmd
Gene Name:
GDP-mannose 4,6-dehydratase
Gene Aliases:
Brazil nut, CG8890, Dm-gmd, DmGMD, Dmel\CG8890, Dmel_CG8890, GMD, gmd, l(2)SH1606, l(2)SH2 1606
Chromosome Location:
Chr.2L: 4975300 - 4977023 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0031661
Assay ID Dm01804543_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gmd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_135044.4 NP_608888.2 2 - 3 700 68
GenBank mRNA BT099628.1 - 2 - 3 666 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

GDP-mannose metabolic process
GDP-L-fucose biosynthetic process
'de novo' GDP-L-fucose biosynthetic process
cell fate commitment
positive regulation of Notch signaling pathway
oogenesis
GDP-mannose 4,6-dehydratase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0